Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB40AL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548322

Catalog No. PA548322


Add to cart

Description

Description

May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications

Specifications

RAB40AL
Polyclonal
PBS with 0.09% sodium azide
P0C0E4
Ras-related protein Rab-40A-like, Ras-like GTPase, RAB40AL, RLGP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
282808
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RAB40AL
Liquid
RAB40AL
KLH conjugated synthetic peptide between 7-36 amino acids from the N-terminal region of human RAB40AL.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit