Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB40C Polyclonal Antibody
RAB40C Polyclonal Antibody
Supplier: Thermo Scientific PA544791
Description
RAB40C Polyclonal Antibody for Western Blot

Specifications
RAB40C | |
Polyclonal | |
Unconjugated | |
RAB40C | |
cb453; rab40c; Rab40c, member RAS oncogene family; RAR (RAS like GTPASE) like; RAR like; RAR3; RARL; rar-like protein; RASL8C; RAS-like GTPase; ras-like protein family member 8C; RAS-like, family 8, member C; ras-related protein Rab-40C; SOCS box containing protein RAR3; SOCS box-containing protein RAR3; Unknown (protein for MGC:136966); wu:fk50d06; zgc:136966 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57799 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96S21 | |
RAB40C | |
Synthetic peptide directed towards the N-terminal of human RAB40C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction