Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570752
Description
RAB43 Polyclonal Antibody for Western Blot
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. The low intrinsic GTPase activity of RAB43 is activated by USP6NL. Involved in retrograde transport from the endocytic pathway to the Golgi apparatus. Involved in the transport of Shiga toxin from early and recycling endosomes to the trans-Golgi network. Required for the structural integrity of the Golgi complex. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.
Specifications
RAB43 | |
Polyclonal | |
Unconjugated | |
RAB43 | |
1810048P08Rik; 2500004H21Rik; AW490415; RAB11B; RAB41; Rab43; RAB43, member RAS oncogene family; ras-related protein Rab-41; Ras-related protein Rab-43 | |
Rabbit | |
Affinity chromatography | |
RUO | |
339122 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86YS6 | |
RAB43 | |
synthetic peptide directed towards the C-terminal region of human RAB43. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction