Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB5B Polyclonal Antibody
RAB5B Polyclonal Antibody
Supplier: Thermo Scientific PA566986
Description
RAB5B Polyclonal Antibody for Western Blot, ICC/IF

Specifications
RAB5B | |
Polyclonal | |
Unconjugated | |
RAB5B | |
C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5869 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P61020 | |
RAB5B | |
Recombinant Human RAB5B. Recombinant protein control fragment (Product #RP-105447). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction