Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB5B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104082
Description
Antibody detects endogenous levels of total RAB5B.
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.
Specifications
RAB5B | |
Polyclonal | |
Unconjugated | |
RAB5B | |
C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19344, 288779, 5869 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61020, P61021 | |
RAB5B | |
A synthesized peptide derived from human RAB5B(Accession P61020), corresponding to amino acid residues E161-Q211. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction