Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab7a Antibody (10E4), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260237MFV610
Description
Rab7a Monoclonal antibody specifically detects Rab7a in Human samples. It is validated for Flow CytometrySpecifications
Rab7a | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
100000 | |
Flow Cytometry | |
CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a | |
Recombinant human RAB7A (1-207aa) purified from E. coli | |
0.1 mL | |
Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction | |
7879 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction