Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab7a Mouse anti-Human, Clone: 100000, Novus Biologicals™

Mouse Monoclonal Antibody

$203.00 - $300.50

Specifications

Antigen Rab7a
Clone 100000
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry 2-5 ug, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60237100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP260237100UL
100 ul Each for $300.50
Add to cart
 
NB6023750UL View Documents Novus BiologicalsSupplier Diversity Partner
NBP26023750UL
50 ul Each for $203.00
Add to cart
 
Description

Description

Rab7a Monoclonal antibody specifically detects Rab7a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Rab7a
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Purified
RUO
Human
CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a
RAB7A
IgG2a Kappa
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
100000
Western Blot 1:1000, Flow Cytometry 2-5 ug, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Unconjugated
Mouse
Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
7879
Recombinant human RAB7A (1-207aa) purified from E. coli
Monoclonal
Protein A purified

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit