Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab7a Rabbit anti-Human, Alexa Fluor 750, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298732AF750
Description
Rab7a Polyclonal specifically detects Rab7a in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Rab7a | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 750 | |
CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Rab7a. | |
0.1 mL | |
Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction | |
7879 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title