Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB8B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen RAB8B
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094348
View Documents
Novus Biologicals
NBP304520100UL
100 μg
Each for $320.71
Add to cart
 
NB094349
View Documents
Novus Biologicals
NBP30452020UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

RAB8B Polyclonal antibody specifically detects RAB8B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

RAB8B
Unconjugated
Rabbit
Signal Transduction
PBS with 50% glycerol, pH7.3.
51762
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
FLJ38125, RAB-8b protein, RAB8B, member RAS oncogene family, ras-related protein Rab-8B
Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human RAB8B (NP_057614.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit