Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB9A Rabbit anti-Human, Clone: SR45-05, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267331
Description
RAB9A Monoclonal antibody specifically detects RAB9A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RAB9A | |
SR45-05 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Purified | |
RAB9A | |
Recombinant Protein | |
Protein A purified | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Signal Transduction | |
Primary | |
9367 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title