Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RABAC1 Recombinant Protein

Supplier:  Abnova™ H00010567P0110

 View more versions of this product

Catalog No. 89-011-969


Explore available promotions
Add to Cart

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

NP_006414
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
45.98
8
Glutathione Sepharose 4 Fast Flow
10 ug
MAAQKDQQKDAEAEGLSGTTLLPKLIPSGAGREWLERRRATIRPWSTFVDQQRFSRPRNLGELCQRLVRNVEYYQSNYVFVFLGLILYCVVTSPMLLVALAVFFGACYILYLRTLESKLVLFGREVSPEHQYALAGGISFPFFWLAGAGSAVFWVLGATLVVIGSHAAFHQIEAVDGEELQMEPV
PRA1/PRAF1/YIP3
RABAC1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10567
RABAC1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RABAC1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.