Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-14-3-3 beta/alpha, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1352541.0ML

Catalog No. NBP13525410


Add to cart

Description

Description

14-3-3 beta/alpha Polyclonal antibody specifically detects 14-3-3 beta/alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

14-3-3 beta/alpha
Unconjugated
PBS (pH 7.6) and 1.0% BSA with 0.1% Sodium Azide
14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha, brain protein 14-3-3, beta isoform, GW128, HS1, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, protein kinase C inhibitor protein-1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alphapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, betapolypeptide, YWHAA
Rabbit
IgG
1 ml
Cancer
Primary
7529
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100
Affinity Purified
YWHAB
A synthetic peptide from N-terminus of human 14-3-3 beta.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit