Learn More
A4GNT Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of A4GNT.
Manufacturer: Abnova Corporation PAB17602
Description
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine (GlcNAc) to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1-->4Galbeta-->R and is largely associated with the Golgi apparatus membrane. [provided by RefSeq
Specifications
A4GNT | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of A4GNT. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q9UNA3 | |
A4GNT | |
A synthetic peptide corresponding to internal of human A4GNT. | |
RUO | |
51146 | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
A4GNT | |
MGC149493/alpha4GnT | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total A4GNT protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only