Learn More
MilliporeSigma™ Rabbit anti-acetyl-Histone H3, Alexa Fluor 488, Polyclonal,
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ 06599AF488
Catalog No. 06-599-AF488
Add to cart
Description
Description
Specifically detects acetyl-Histone H3 in Human, Mouse samples, and it is validated for Immunocytochemistry
Histone H3 is one of the five main histone proteins that are involved in the structure of chromatin in eukaryotic cells. The chromatin fiber is composed on repetitive units, known as nucleosomes, which are comprised of an octamer of core histones (two each of H2A, H2B, H3, and H4), around which DNA are wrapped. Histone H3 features a main globular domain and a long N-terminal tail. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 is specifically phosphorylated during both mitosis and meiosis in patterns that are specifically coordinated in space and time. These phosphorylations are initiated at different phases of the cell division. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.
Specifications
Specifications
acetyl-Histone H3
Alexa Fluor 488
Q16695
HIST3H3; H3FT
Synthetic peptide corresponding to human histone H3 N-terminal sequence with acetylated Lys9 and Lys14
100μL
Epigenetics and Nuclear Function
Primary
NP_003484.1
Immunocytochemistry
Purified rabbit antibody conjugate in buffer containing PBS, 15mg/mL BSA and 0.1% Sodium Azide.
Purified
Rabbit
Protein A Purfied
RUO
Stable for one year at 2°C to 8°C from date of receipt.
Polyclonal
Human, Murine