Learn More
AKT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKT1.
Supplier:  Abnova Corporation PAB4548
Catalog No. 89-131-460
Add to Cart
Description
Description
The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Specifications
Specifications
AKT1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of AKT1.
In PBS (0.09% sodium azide)
P31749
AKT1
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human AKT1.
400 μL
Primary
Human
Liquid
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user.
AKT1
AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA
Rabbit
Protein G purification
RUO
207
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions