Learn More
AKT2 (phospho S474), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT2.
Supplier: Abnova Corporation PAB8183
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Specifications
AKT2 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
AKT2 | |
AKT2 | |
Synthetic phosphopeptide corresponding to residues surrounding S474 of human AKT2. | |
100 μL | |
Primary | |
Akt2 (phospho-Ser474) antibody detects endogenous levels of Akt2 only when phosphorylated at serine 474. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of AKT2. | |
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
PKBB/PKBBETA/PRKBB/RAC-BETA | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
208 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only