Learn More
Alb Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Alb.
Supplier:  Abnova Corporation PAB5079
Catalog No. 89-131-726
Add to Cart
Description
Description
Specifications
Specifications
Alb
Polyclonal
Rabbit polyclonal antibody raised against native Alb.
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Alb1/Albza
Rabbit
150 μg
Primary
Rat
Liquid
ELISA, Immunoprecipitation, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Alb
Alb
Native purified rat Alb.
RUO
24186
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions