Learn More
ALDH8A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDH8A1.
Supplier: Abnova Corporation PAB3143
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
ALDH8A1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ALDH8A1. | |
In PBS (0.09% sodium azide) | |
NP_072090;Q9H2A2 | |
ALDH8A1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ALDH8A1. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
ALDH8A1 | |
ALDH12/DJ352A20.2/DKFZp779D2315/MGC138650 | |
Rabbit | |
Protein A purification | |
RUO | |
64577 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only