Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: P16-D, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1301490.5ML

 View more versions of this product

Catalog No. NBP13014905


Add to cart

Description

Description

Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Alkaline Phosphatase/ALPP
P16-D
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-200, Immunohistochemistry-Frozen 1:100
P05187
Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Rabbit
IgG
0.5 ml
Embryonic Stem Cell Markers, Stem Cell Markers
Primary
250
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
ALPP
Peptide derived from the middle portion of human PLAP sequence.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This antibody is specific to an internal portion of human PLAP sequence.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only