Learn More
Rabbit anti-AMELX/AMELY, Polyclonal, Abnova™
Manufacturer:  Abnova Corporation PAB13562
Catalog No. 89-116-742
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
AMELX/AMELY
Unconjugated
Liquid
Rabbit
100μL
Primary
281620;281621
Immunohistochemistry (Paraffin), Western Blot
In serum
AMELX
Native purified AMELX/AMELY from bovine amelogenin .
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Bovine, Human, Murine, Rat