Learn More
Amelx Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Amelx.
Supplier:  Abnova Corporation PAB5867
Catalog No. 89-131-961
Add to Cart
Description
Description
Specifications
Specifications
Amelx
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of Amelx.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
ALGN/AMGL/AMGX/Amel/Amg
Rabbit
100 μg
Primary
Mouse, Rat
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Amelx
Amelx
A synthetic peptide corresponding to N-terminus of mouse Amelx.
RUO
11704
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions