Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amy1a, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Amy1a.
Supplier: Abnova Corporation PAB17955
Specifications
Amy1a | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Amy1a. | |
In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
NP_001010970 | |
Amy1a | |
A synthetic peptide corresponding to the residues surrounding amino acid 273 of rat Amy1a. | |
RUO | |
24203 | |
Chicken, Human, Mouse, Pig, Rat | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
Amy1a | |
Amy1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼58 KDa alpha-Amylase in samples from human, mouse and rat origins. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction