Learn More
Rabbit anti-ANGPTL1, Polyclonal, Abnova™
Manufacturer: Abnova Corporation PAB18288
Specifications
ANGPTL1 | |
Polyclonal | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ANGPTL1 | |
A synthetic peptide corresponding to human ANGPTL1. | |
100 μg | |
9068 |