Learn More
ANKH Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANKH.
Supplier:  Abnova Corporation PAB5241
Catalog No. 89-131-780
Add to Cart
Description
Description
This gene encodes a multipass transmembrane protein that is expressed in joints and other tissues and controls pyrophosphate levels in cultured cells. Progressive ankylosis-mediated control of pyrophosphate levels has been suggested as a possible mechanism regulating tissue calcification and susceptibility to arthritis in higher animals. Mutations in this gene have been associated with autosomal dominant craniometaphyseal dysplasia. [provided by RefSeq
Specifications
Specifications
ANKH
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of ANKH.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
ANK/CCAL2/CMDJ/CPPDD/FLJ27166/HANK/MANK
Rabbit
100 μg
Primary
Human, Mouse
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
ANKH
ANKH
A synthetic peptide corresponding to internal region of human ANKH.
RUO
56172
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions