Learn More
ANXA2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANXA2.
Manufacturer: Abnova Corporation PAB19088
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
ANXA2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ANXA2. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
ANX2/ANX2L4/CAL1H/LIP2/LPC2/LPC2D/P36/PAP-IV | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
302 | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) Immunohistochemistry (Frozen sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
ANXA2 | |
ANXA2 | |
A synthetic peptide corresponding to internal region of human ANXA2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only