Learn More
ANXA3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant ANXA3.
Manufacturer: Abnova Corporation PAB19129
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation. [provided by RefSeq
Specifications
ANXA3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against full length recombinant ANXA3. | |
In serum | |
P12429 | |
ANXA3 | |
Recombinant protein corresponding to full length human ANXA3. | |
RUO | |
306 | |
Human |
ELISA, Western Blot | |
Unconjugated | |
Western blot (1:500) The optimal working dilution should be determined by the end user. | |
ANXA3 | |
ANX3 | |
Rabbit | |
100 μL | |
Primary | |
It can expression in BGC823 whole cell lysate. | |
Liquid |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only