Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AOF2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of AOF2.

Manufacturer:  Abnova Corporation PAB14121

Catalog No. 89-116-941


Add to cart

Description

Description

This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Specifications

AOF2
Unconjugated
ELISA (1:200-1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
AOF2
Liquid
AOF2
A synthetic peptide (conjugated with KLH) corresponding to human AOF2.
RUO
Primary
23028
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of AOF2.
In PBS (0.05% sodium azide, 0.05% proclin 300)
O60341
BHC110|KDM1|KIAA0601|LSD1
Rabbit
50 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only