Learn More
APH1A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
Supplier:  Abnova Corporation PAB4887
Catalog No. 89-131-633
Add to Cart
Description
Description
APH1 is a multipass transmembrane protein that interacts with presenilin (see PSEN1; MIM 104311) and nicastrin (APH2; MIM 605254) as a functional component of the gamma-secretase complex. The gamma-secretase complex is required for the intramembrane proteolysis of a number of membrane proteins, including the amyloid-beta precursor protein (APP; MIM 104760) and Notch (MIM 190198).[supplied by OMIM
Specifications
Specifications
APH1A
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
In PBS (0.09% sodium azide)
NP_001071096;NP_057106;Q96BI3
APH1A
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human APH1A.
400 μL
Primary
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
APH1A
6530402N02Rik/APH-1A/CGI-78
Rabbit
Protein G purification
RUO
51107
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations