Learn More
APH1A Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
Manufacturer:  Abnova Corporation PAB13237
Catalog No. 89-116-484
Add to cart
Description
Description
APH1 is a multipass transmembrane protein that interacts with presenilin (see PSEN1; MIM 104311) and nicastrin (APH2; MIM 605254) as a functional component of the gamma-secretase complex. The gamma-secretase complex is required for the intramembrane proteolysis of a number of membrane proteins, including the amyloid-beta precursor protein (APP; MIM 104760) and Notch (MIM 190198).[supplied by OMIM
Specifications
Specifications
APH1A
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
APH1A
Liquid
APH1A
A synthetic peptide corresponding to N-terminus 13 amino acids of human APH1A.
RUO
Primary
51107
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
In PBS (0.02% sodium azide)
AAH08732
6530402N02Rik|APH-1A|CGI-78
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine