Learn More
APLNR Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of APLNR.
Manufacturer: Abnova Corporation PAB25525
Description
This gene encodes a member of the G protein-coupled receptor gene family. The encoded protein is related to the angiotensin receptor, but is actually an apelin receptor that inhibits adenylate cyclase activity and plays a counter-regulatory role against the pressure action of angiotensin II by exerting hypertensive effect. It functions in the cardiovascular and central nervous systems, in glucose metabolism, in embryonic and tumor angiogenesis and as a human immunodeficiency virus (HIV-1) coreceptor. Two transcript variants resulting from alternative splicing have been identified. [provided by RefSeq
Specifications
APLNR | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7-14 ug/mL) The optimal working dilution should be determined by the end user. | |
APLNR | |
AGTRL1|APJ|APJR|FLJ90771|MGC45246 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
187 | |
Human |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of APLNR. | |
In PBS (0.09% sodium azide) | |
Liquid | |
APLNR | |
A synthetic peptide corresponding to 20 amino acids at 3rd cytoplasmic domain of human APLNR. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only