Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apod, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Apod.
Supplier: Abnova Corporation PAB5870
Specifications
Apod | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Apod. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
Apod | |
A synthetic peptide corresponding to N-terminus of mouse Apod. | |
RUO | |
11815 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Apod | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only