Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqp4, Rabbit, PerCP, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant Rat Aqp4.

Supplier:  Abnova Corporation PAB28907

Catalog No. 89-130-433


Add to Cart

Description

Description

Sequence: CTKGSYMEVEDNRSQVETED
Specifications

Specifications

Aqp4
Polyclonal
Rabbit polyclonal antibody raised against recombinant Rat Aqp4.
PBS pH 7.4, 50% glycerol and 0.09% sodium azide
NP_036957.1
Aqp4
Recombinant protein corresponding to Rat Aqp4 C-terminus.
100 μg
Primary
0.5 ug/mL of PAB28907 was sufficient for detection of Aqp4, ∼35kDa and sometimes larger glycosylated bands ∼50kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Immunofluorescence, Western Blot
PerCP
Immunofluorescence (1:400) Western Blot (1:1000-4000) The optimal working dilution should be determined by the end user.
Aqp4
AQP-4/AQP4.M23/MIWC2/Miwc
Rabbit
Affinity purification
RUO
25293
Human, Mouse, Rat
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only