Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARG1 Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against native ARG1 (Bovine).

Supplier:  Abnova Corporation PAB28072

Catalog No. 89-129-581


Add to Cart

Description

Description

Specifications

Specifications

ARG1
Polyclonal
Rabbit polyclonal antibody raised against native ARG1 (Bovine).
Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
MGC128755
Rabbit
100 μg
Primary
Bovine
Lyophilized
ELISA, Western Blot
Unconjugated
ELISA (1:5000 - 1:25000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
ARG1
ARG1
Native purified ARG1 from bovine liver.
RUO
513608
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only