Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243545
Description
Specifically detects ARHGAP17 in Mouse samples, and it is validated for Western Blotting
ARHGAP17 Polyclonal specifically detects ARHGAP17 in Mouse samples. It is validated for Western Blot.Specifications
ARHGAP17 | |
Polyclonal | |
Western Blot 1:500-1:3000 | |
DKFZp564A1363, FLJ10308, FLJ13219, FLJ37567, FLJ43368, MGC87805, MST110, MSTP038, MSTP066, MSTP110, NADRIN, neuron-associated developmentally regulated protein, PP367, PP4534, Rho GTPase activating protein 17, rho GTPase-activating protein 17, RhoGAP interacting with CIP4 homologs 1, RhoGAP interacting with CIP4 homologs protein 1, Rho-type GTPase-activating protein 17, RICH-1, RICH1B, RICH1MST066, WBP15 | |
Rabbit | |
95 kDa | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
ARHGAP17 | |
Recombinant protein encompassing a sequence within the center region of human ARHGAP17. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
55114 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction