Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-ASMT, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243801

Catalog No. NBP243801


Add to cart

Description

Description

Specifically detects ASMT in Human samples, and it is validated for Western Blotting

ASMT Polyclonal antibody specifically detects ASMT in Human samples. It is validated for Western Blot.
Specifications

Specifications

ASMT
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
acetylserotonin N-methyltransferase, acetylserotonin O-methyltransferaseEC 2.1.1.4, ASMTY, HIOMTacetylserotonin methyltransferase (Y chromosome), HIOMTY, hydroxyindole O-methyltransferase
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
438
Western Blot
Western Blot 1:500-1:3000
Ion Exchange Chromatography
ASMT
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ASMT. The exact sequence is proprietary.
38 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only