Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF7, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of ATF7.

Supplier:  Abnova Corporation PAB27155

Catalog No. 89-128-808


Add to Cart

Description

Description

Specifications

Specifications

ATF7
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
ATF7
ATFA/MGC57182
Rabbit
Antigen affinity purification
RUO
11016
Human, Mouse
Liquid
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of ATF7.
In PBS, pH 7.2 (0.05% sodium azide)
P17544
ATF7
A synthetic peptide corresponding to human ATF7.
100 μL
Primary
ATF7 polyclonal antibody detects endogenous levels of ATF7 protein.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only