Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-ATG14, synthetic peptide, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243644

Catalog No. NBP243644


Add to cart

Description

Description

Specifically detects ATG14 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

ATG14 Polyclonal antibody specifically detects ATG14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ATG14
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
22863
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Ion Exchange Chromatography
ATG14
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ATG14. The exact sequence is proprietary.
55 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only