Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Atpalpha, (aa 578-839), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242817

Catalog No. NBP242817


Add to cart

Description

Description

Specifically detects Atpalpha in Drosophila, Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Atpalpha Polyclonal antibody specifically detects Atpalpha in Human, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Atpalpha
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Drosophila
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1:5000-1:20000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
48971
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only