Learn More
AURKC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AURKC.
Supplier: Abnova Corporation PAB4512
Description
This gene encodes a member of the Aurora subfamily of serine/threonine protein kinases. The encoded protein is a chromosomal passenger protein that forms complexes with Aurora-B and inner centromere proteins and may play a role in organizing microtubules in relation to centrosome/spindle function during mitosis. This gene is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
AURKC | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of AURKC. | |
In PBS (0.09% sodium azide) | |
Q9UQB9 | |
AURKC | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human AURKC. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
AURKC | |
AIE2/AIK3/AurC/STK13/aurora-C | |
Rabbit | |
Protein G purification | |
RUO | |
6795 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only