Learn More
Rabbit anti-Aurora A, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2367370.1MG
Catalog No. NBP23673701
Add to cart
Description
Description
Aurora A Polyclonal antibody specifically detects Aurora A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
Specifications
Aurora A
0.5 mg/mL
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
O14965
AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7
Rabbit
IgG
0.1 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors
Primary
6790
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
PBS containing 0.05% BSA with 0.05% Sodium Azide
Purified
AURKA
A portion of amino acids 100-150 of human Aurora was used as the immunogen for this antibody.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat