Learn More
AVEN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AVEN.
Manufacturer:  Abnova Corporation PAB13091
Catalog No. 89-116-343
Add to cart
Description
Description
Specifications
Specifications
AVEN
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
AVEN
Liquid
AVEN
A synthetic peptide corresponding to C-terminus of human AVEN.
RUO
Primary
57099
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of AVEN.
In PBS (0.02% sodium azide)
NP_065104
PDCD12
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat