Learn More
AXIN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Lytechinus variegatus AXIN.
Manufacturer:  Abnova Corporation PAB16080
Catalog No. 89-138-163
Add to cart
Description
Description
Specifications
Specifications
AXIN
Unconjugated
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to C-terminus of Lytechinus variegatus AXIN.
RUO
Primary
Sea urchin
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Lytechinus variegatus AXIN.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal