Learn More
BAIAP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BAIAP2.
Supplier:  Abnova Corporation PAB4062
Catalog No. 89-131-257
Add to Cart
Description
Description
The protein encoded by this gene has been identified as a brain-specific angiogenesis inhibitor (BAI1)-binding protein. This adaptor protein links membrane bound G-proteins to cytoplasmic effector proteins. This protein functions as an insulin receptor tyrosine kinase substrate and suggests a role for insulin in the central nervous system. It also associates with a downstream effector of Rho small G proteins, which is associated with the formation of stress fibers and cytokinesis. This protein is involved in lamellipodia and filopodia formation in motile cells and may affect neuronal growth-cone guidance. This protein has also been identified as interacting with the dentatorubral-pallidoluysian atrophy gene, which is associated with an autosomal dominant neurodegenerative disease. Alternative splicing results in multiple transcript variants encoding distinct isoforms
Specifications
Specifications
BAIAP2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of BAIAP2.
In PBS (0.09% sodium azide)
NP_006331
BAIAP2
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human BAIAP2.
400 μL
Primary
Human, Mouse
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
BAIAP2
BAP2/IRSP53
Rabbit
Protein G purification
RUO
10458
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions