Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCHE, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant BCHE.

Supplier:  Abnova Corporation PAB28449

Catalog No. 89-129-955


Add to Cart

Description

Description

Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage. [provided by RefSeq

Sequence: QQLALQWVQKNIAAFGGNPKSVTLFGESAGAASVSLHLLSPGSHSLFTRAILQSGSFNAPWAVTSLYEARNRTLNLAKLTGCSRENETEIIKCLRNKDPQEILLNEAFVVPYGTPLSVNFGPTVD
Specifications

Specifications

BCHE
Polyclonal
Rabbit polyclonal antibody raised against recombinant BCHE.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
F8WEX7
BCHE
Recombinant protein corresponding to amino acids of recombinant BCHE.
100 μL
Primary
Human
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry(1:50-1:200) The optimal working dilution should be determined by the end user.
BCHE
CHE1/E1
Rabbit
Antigen affinity purification
RUO
590
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only