Learn More
Rabbit anti-Bcl-2, Clone: P21-A, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179011100UL
 View more versions of this product
Catalog No. NBP17901110
Add to cart
Description
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Bcl-2
P21-A
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200
P10415
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Rabbit
IgG
100 ul
Apoptosis, Autophagy, Cancer, Cellular Markers, Tumor Suppressors
Primary
596
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
BCL2
peptide derived from the sequence close to N-terminal region of human bcl-2
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human