Learn More
BCL2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BCL2.
Manufacturer:  Abnova Corporation PAB13118
Catalog No. 89-116-370
Add to cart
Description
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Specifications
Specifications
BCL2
Unconjugated
Western Blot (1-2 ug/mL) Immunohistochemistry (2 ug/mL) The optimal working dilution should be determined by the end user.
BCL2
Liquid
BCL2
A synthetic peptide corresponding to N-terminus 15 amino acids of human BCL2.
RUO
Primary
596
Human, Murine
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of BCL2.
In PBS (0.02% sodium azide)
AAH27258
Bcl-2
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
It is human and mouse reactive.