Learn More
BCR (phospho Y177), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BCR.
Manufacturer: Abnova Corporation PAB25329
Description
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: P-F-Yp-V-NSpecifications
BCR | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
BCR | |
Liquid | |
BCR | |
Synthetic phosphopeptide corresponding to residues surrounding Y177 of human BCR. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine |
Immunohistochemistry (PFA fixed), Western Blot | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BCR. | |
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P11274 | |
ALL|BCR-ABL1|BCR1|CML|D22S11|D22S662|FLJ16453|PHL | |
Rabbit | |
Affinity purification | |
RUO | |
Primary | |
613 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only