Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BECN1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against BECN1.

Manufacturer:  Abnova Corporation PAB12474

Catalog No. 89-115-849


Add to cart

Description

Description

Specifications

Specifications

BECN1
Unconjugated
Western Blot (1:500) The optimal working dilution should be determined by the end user.
BECN1
Liquid
BECN1
Full length human BECN1.
RUO
Primary
8678
Human, Murine
Western Blot
Rabbit polyclonal antibody raised against BECN1.
In antiserum (0.09% sodium azide)
Q14457
ATG6|VPS30|beclin1
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody is specific to Beclin 1.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only