Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Amyloid 42 Rabbit anti-Human, Mouse, Rat, Canine, Guinea Pig, Primate, Rabbit, Sheep, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244113

Catalog No. NBP244113


Add to cart

Description

Description

Specifically detects beta Amyloid 42 in Human, Mouse, Primate, Rat samples, and it is validated for ELISA, Western Blotting

beta Amyloid 42 Polyclonal antibody specifically detects beta Amyloid 42 in Human, Mouse, Rat, Canine, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

beta Amyloid 42
1.0 mg/mL
Western Blot 1 ug/ml, ELISA 1: 10000, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified
Anti-Beta Amyloid 42 affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminus of AB42.
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Canine, Guinea Pig, Primate, Rabbit, Sheep
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Rabbit
IgG
0.05 mg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only