Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC5 (phospho T34), Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.

Manufacturer:  Abnova Corporation PAB12585

Catalog No. 89-115-929


Add to cart

Description

Description

This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq

Specifications

Specifications

BIRC5
Unconjugated
Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
BIRC5
API4|EPR-1
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody is specific to phosphorylated survivin.
Immunocytochemistry, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Liquid
BIRC5
Synthetic phosphopeptide corresponding to residues surrounding T34 of BIRC5.
RUO
Primary
332
Bovine, Canine, Feline, Human, Murine, Porcine, Primate
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only